Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYP73 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203224.2 | NP_001190153.1 | 1 - 2 | 259 | 78 | |
NM_001340100.1 | NP_001326526.1 | 1 - 2 | 246 | 78 | ||
NM_001340101.1 | NP_001326527.1 | 1 - 2 | 246 | 78 | ||
NM_001340103.1 | NP_001326529.1 | 1 - 2 | 228 | 78 | ||
NM_116010.1 | NP_567114.1 | 1 - 2 | 218 | 78 | ||
GenBank mRNA | AF355759.1 | - | 1 - 2 | 218 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |