Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LHCA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116012.5 | NP_191706.2 | 2 - 3 | 469 | 74 | |
GenBank mRNA | AF134120.1 | - | 2 - 3 | 285 | 74 | |
AY054670.1 | - | 2 - 3 | 287 | 74 | ||
AY065429.1 | - | 2 - 3 | 314 | 74 | ||
AY072483.1 | - | 2 - 3 | 247 | 74 | ||
BT002070.1 | - | 2 - 3 | 287 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |