Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RS31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116051.4 | NP_567120.1 | 3 - 4 | 367 | 113 | |
GenBank mRNA | AF439831.1 | - | 3 - 4 | 306 | 113 | |
AY125565.1 | - | 2 - 3 | 222 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |