Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATMYC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001160722.2 | NP_001154194.1 | 7 - 8 | 1602 | 75 | |
NM_116272.4 | NP_191957.2 | 7 - 8 | 1747 | 75 | ||
GenBank mRNA | AB493664.1 | - | 7 - 8 | 1505 | 75 | |
AF251697.1 | - | 7 - 8 | 1505 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |