Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: DFL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340446.1 | NP_001319858.1 | 3 - 4 | 1916 | 144 | |
NM_116578.3 | NP_192249.1 | 1 - 2 | 684 | 144 | ||
GenBank mRNA | AY059941.1 | - | 1 - 2 | 448 | 144 | |
BT000061.1 | - | 1 - 2 | 313 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |