Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MED31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203413.1 | NP_001190342.1 | 3 - 4 | 414 | 93 | |
NM_121998.5 | NP_197491.2 | 3 - 4 | 332 | 93 | ||
GenBank mRNA | AY072222.1 | - | 3 - 4 | 284 | 93 | |
BT002744.1 | - | 3 - 4 | 245 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |