Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FRS6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104130.4 | NP_175661.1 | 1 - 2 | 361 | 86 | |
GenBank mRNA | AY080775.1 | - | 1 - 2 | 193 | 86 | |
AY133776.1 | - | 1 - 2 | 29 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |