Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XIE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333655.1 | NP_001320715.1 | 36 - 37 | 4738 | 131 | |
NM_104334.2 | NP_175858.1 | 35 - 36 | 4462 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |