Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLAH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104908.4 | NP_176418.2 | 1 - 2 | 928 | 106 | |
GenBank mRNA | BT021090.1 | - | 1 - 2 | 862 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |