Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ESD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341077.1 | NP_001328371.1 | 8 - 9 | 1644 | 129 | |
NM_117680.4 | NP_567478.1 | 8 - 9 | 1607 | 129 | ||
GenBank mRNA | AF386934.1 | - | 8 - 9 | 1532 | 129 | |
AY081548.1 | - | 8 - 9 | 1408 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |