Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036843.1 | NP_001031920.1 | 5 - 6 | 1031 | 161 | |
NM_001203428.1 | NP_001190357.1 | 5 - 6 | 1067 | 161 | ||
NM_001343706.1 | NP_001331520.1 | 5 - 6 | 1054 | 161 | ||
NM_147876.5 | NP_680181.2 | 5 - 6 | 1054 | 161 | ||
GenBank mRNA | AK316941.1 | - | 5 - 6 | 965 | 161 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |