Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105445.2 | NP_176943.1 | 3 - 4 | 1351 | 86 | |
GenBank mRNA | DQ446408.1 | - | 3 - 4 | 1172 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |