Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PUM23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334523.1 | NP_001321803.1 | 8 - 9 | 1859 | 114 | |
NM_105891.3 | NP_177376.1 | 8 - 9 | 1709 | 114 | ||
NM_179547.5 | NP_849878.2 | 8 - 9 | 1644 | 114 | ||
NM_202401.1 | NP_974130.1 | 8 - 9 | 1614 | 114 | ||
GenBank mRNA | AY035041.1 | - | 8 - 9 | 1651 | 114 | |
AY051073.1 | - | 7 - 8 | 1540 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |