Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_114147.5 | NP_189865.1 | 1 - 2 | 272 | 64 | |
GenBank mRNA | AY062851.1 | - | 1 - 2 | 237 | 64 | |
AY114574.1 | - | 1 - 2 | 98 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |