Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: TIF3H1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100960.3 | NP_563880.1 | 5 - 6 | 693 | 102 | |
NM_202079.1 | NP_973808.1 | 4 - 5 | 579 | 102 | ||
GenBank mRNA | AY054641.1 | - | 5 - 6 | 527 | 102 | |
AY081546.1 | - | 4 - 5 | 376 | 102 | ||
AY087338.1 | - | 5 - 6 | 595 | 102 | ||
BT000765.1 | - | 5 - 6 | 472 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |