Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: SPDS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102230.5 | NP_173794.3 | 6 - 7 | 619 | 95 | |
NM_202171.2 | NP_973900.2 | 6 - 7 | 764 | 95 | ||
GenBank mRNA | AJ251296.1 | - | 6 - 7 | 676 | 95 | |
AY062593.1 | - | 6 - 7 | 687 | 95 | ||
AY093360.1 | - | 6 - 7 | 619 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |