Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: NHX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337650.1 | NP_001327378.1 | 4 - 5 | 589 | 151 | |
NM_001337651.1 | NP_001327379.1 | 4 - 5 | 589 | 151 | ||
NM_001337652.1 | NP_001327377.1 | 3 - 4 | 519 | 151 | ||
NM_111512.3 | NP_187288.2 | 4 - 5 | 589 | 151 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |