Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118536.4 | NP_194135.1 | 8 - 9 | 1685 | 88 | |
GenBank mRNA | AK118407.1 | - | 8 - 9 | 1542 | 88 | |
BT010732.1 | - | 8 - 9 | 1483 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |