Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HVA22D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001125576.1 | NP_001119048.1 | 2 - 3 | 632 | 69 | |
NM_118628.4 | NP_567713.1 | 3 - 4 | 585 | 69 | ||
GenBank mRNA | AF141662.1 | - | 3 - 4 | 504 | 69 | |
AY059750.1 | - | 3 - 4 | 504 | 69 | ||
AY086850.1 | - | 3 - 4 | 504 | 69 | ||
AY113978.1 | - | 3 - 4 | 381 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |