Search Thermo Fisher Scientific
Gene Symbol: NAP1;1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001125588.2 | NP_001119060.1 | 3 - 4 | 351 | 61 | |
NM_118744.3 | NP_194341.1 | 3 - 4 | 328 | 61 | ||
GenBank mRNA | AY054484.1 | - | 3 - 4 | 245 | 61 | |
AY093283.1 | - | 3 - 4 | 184 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |