Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PFK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118760.3 | NP_567742.1 | 5 - 6 | 796 | 139 | |
GenBank mRNA | AY049245.1 | - | 5 - 6 | 704 | 139 | |
AY090267.1 | - | 5 - 6 | 386 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |