Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341838.1 | NP_001328096.1 | 3 - 4 | 574 | 96 | |
NM_118852.3 | NP_567768.1 | 4 - 5 | 757 | 96 | ||
GenBank mRNA | D21137.1 | - | 4 - 5 | 601 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |