Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341859.1 | NP_001328990.1 | 3 - 4 | 746 | 91 | |
NM_118885.4 | NP_194480.2 | 3 - 4 | 836 | 91 | ||
GenBank mRNA | AJ002020.2 | - | 3 - 4 | 655 | 91 | |
AY050365.1 | - | 3 - 4 | 675 | 91 | ||
AY143914.1 | - | 2 - 3 | 550 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |