Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337990.1 | NP_001326831.1 | 1 - 2 | 594 | 157 | |
NM_112071.3 | NP_187842.1 | 2 - 3 | 672 | 157 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |