Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CKX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119120.2 | NP_194703.1 | 3 - 4 | 1175 | 107 | |
NM_179139.1 | NP_849470.1 | 3 - 4 | 1093 | 107 | ||
GenBank mRNA | AY054460.1 | - | 3 - 4 | 1095 | 107 | |
BT000179.1 | - | 3 - 4 | 1007 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |