Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EDM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203616.2 | NP_001190545.1 | 11 - 12 | 2081 | 101 | |
NM_001345123.1 | NP_001332152.1 | 10 - 11 | 2349 | 101 | ||
NM_124921.5 | NP_200350.2 | 11 - 12 | 2136 | 101 | ||
GenBank mRNA | KF112069.1 | - | 10 - 11 | 1958 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |