Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BGAL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119799.4 | NP_568001.1 | 1 - 2 | 510 | 69 | |
NM_179175.2 | NP_849506.1 | 1 - 2 | 305 | 69 | ||
GenBank mRNA | AJ270299.1 | - | 1 - 2 | 227 | 69 | |
AY056285.1 | - | 1 - 2 | 286 | 69 | ||
AY091432.1 | - | 1 - 2 | 187 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |