Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342124.1 | NP_001329752.1 | 13 - 14 | 2189 | 93 | |
NM_119353.4 | NP_194929.2 | 15 - 16 | 2549 | 93 | ||
GenBank mRNA | AB206554.1 | - | 11 - 12 | 1735 | 93 | |
AK228013.1 | - | 15 - 16 | 2392 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |