Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036707.1 | NP_001031784.1 | 9 - 10 | 899 | 67 | |
NM_001342222.1 | NP_001328574.1 | 8 - 9 | 913 | 67 | ||
NM_119514.4 | NP_567928.1 | 9 - 10 | 1005 | 67 | ||
GenBank mRNA | AK221870.1 | - | 9 - 10 | 897 | 67 | |
AY034930.1 | - | 8 - 9 | 760 | 67 | ||
AY088420.1 | - | 8 - 9 | 797 | 67 | ||
AY122986.1 | - | 8 - 9 | 705 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |