Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RKP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202644.1 | NP_001189573.1 | 3 - 4 | 2747 | 149 | |
NM_179689.3 | NP_850020.1 | 3 - 4 | 2734 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |