Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VHA-A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_179791.2 | NP_850122.1 | 15 - 16 | 2225 | 72 | |
GenBank mRNA | AY091008.1 | - | 15 - 16 | 2129 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |