Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHT4;1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336220.1 | NP_001325344.1 | 4 - 5 | 1173 | 111 | |
NM_128519.6 | NP_180526.1 | 5 - 6 | 1126 | 111 | ||
NM_179805.2 | NP_850136.1 | 5 - 6 | 1141 | 111 | ||
NM_201832.1 | NP_973561.1 | 5 - 6 | 939 | 111 | ||
GenBank mRNA | BT001983.1 | - | 5 - 6 | 1100 | 111 | |
BT008489.1 | - | 5 - 6 | 1052 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |