Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOM40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338439.1 | NP_001326561.1 | 4 - 5 | 816 | 123 | |
NM_112890.5 | NP_188634.1 | 4 - 5 | 624 | 123 | ||
GenBank mRNA | AY050925.1 | - | 4 - 5 | 466 | 123 | |
AY117216.1 | - | 4 - 5 | 401 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |