Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CPK9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338479.1 | NP_001327803.1 | 7 - 8 | 1651 | 104 | |
NM_112932.3 | NP_188676.1 | 6 - 7 | 2013 | 104 | ||
GenBank mRNA | AY120727.1 | - | 6 - 7 | 1631 | 104 | |
BT008898.1 | - | 6 - 7 | 1444 | 104 | ||
U31751.1 | - | 6 - 7 | 1518 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |