Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSL03 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202724.2 | NP_001189653.1 | 22 - 23 | 3174 | 65 | |
NM_001336361.1 | NP_001318331.1 | 23 - 24 | 2886 | 65 | ||
NM_001336362.1 | NP_001323730.1 | 23 - 24 | 2640 | 65 | ||
NM_001336363.1 | NP_001323731.1 | 23 - 24 | 2811 | 65 | ||
NM_179847.3 | NP_850178.2 | 22 - 23 | 2814 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |