Search Thermo Fisher Scientific
Gene Symbol: ACX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100511.3 | NP_172119.1 | 1 - 2 | 678 | 80 | |
GenBank mRNA | AF253474.1 | - | 1 - 2 | 516 | 80 | |
AK176257.1 | - | 1 - 2 | 534 | 80 | ||
AY099579.1 | - | 1 - 2 | 534 | 80 | ||
BT008413.1 | - | 1 - 2 | 463 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |