Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AT2G45360 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130099.2 | NP_182061.1 | 1 - 2 | 88 | 67 | |
GenBank mRNA | AK118397.1 | - | 1 - 2 | 88 | 67 | |
BT005233.1 | - | 1 - 2 | 61 | 67 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |