Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337302.1 | NP_001324284.1 | 4 - 5 | 717 | 71 | |
NM_130382.2 | NP_566128.1 | 5 - 6 | 617 | 71 | ||
GenBank mRNA | AK118448.1 | - | 5 - 6 | 538 | 71 | |
AY088692.1 | - | 5 - 6 | 561 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |