Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: U2A' | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100848.4 | NP_172447.1 | 2 - 3 | 216 | 101 | |
GenBank mRNA | AY059902.1 | - | 2 - 3 | 179 | 101 | |
BT000143.1 | - | 2 - 3 | 127 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |