Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100971.3 | NP_172565.1 | 1 | 332 | 63 | |
GenBank mRNA | AY086012.1 | - | 1 | 139 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |