Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: GS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125126.3 | NP_568858.1 | 2 - 3 | 427 | 106 | |
GenBank mRNA | AY063967.1 | - | 2 - 3 | 335 | 106 | |
AY084531.1 | - | 2 - 3 | 335 | 106 | ||
AY114031.1 | - | 2 - 3 | 306 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |