Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CER3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125164.3 | NP_200588.2 | 3 - 4 | 932 | 80 | |
GenBank mRNA | AB099512.1 | - | 3 - 4 | 904 | 80 | |
AB116548.1 | - | 3 - 4 | 613 | 80 | ||
AY131334.1 | - | 3 - 4 | 903 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |