Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRH75 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125613.3 | NP_201025.1 | 6 - 7 | 1212 | 100 | |
GenBank mRNA | AY056137.1 | - | 6 - 7 | 1138 | 100 | |
BT008581.1 | - | 6 - 7 | 1047 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |