Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PAB8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198255.1 | NP_001185184.1 | 1 - 2 | 565 | 86 | |
NM_103863.3 | NP_564554.1 | 1 - 2 | 564 | 86 | ||
GenBank mRNA | AF370517.1 | - | 1 - 2 | 559 | 86 | |
AY064622.1 | - | 1 - 2 | 299 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |