Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343120.1 | NP_001332648.1 | 2 - 3 | 748 | 73 | |
NM_121090.4 | NP_568231.1 | 2 - 3 | 723 | 73 | ||
GenBank mRNA | AY150507.1 | - | 1 - 2 | 462 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |