Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PKL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335976.1 | NP_001324292.1 | 27 - 28 | 3295 | 119 | |
NM_001335977.1 | NP_001324293.1 | 27 - 28 | 3322 | 119 | ||
NM_001335978.1 | NP_001324291.1 | 26 - 27 | 3244 | 119 | ||
NM_128074.4 | NP_565587.1 | 27 - 28 | 3268 | 119 | ||
GenBank mRNA | AF185577.1 | - | 27 - 28 | 3268 | 119 | |
AF185578.1 | - | 26 - 27 | 3248 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |