Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: RCY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084493.1 | NP_001077962.1 | 6 - 7 | 931 | 99 | |
NM_001084494.1 | NP_001077963.1 | 5 - 6 | 1022 | 99 | ||
NM_128200.5 | NP_565622.1 | 6 - 7 | 961 | 99 | ||
GenBank mRNA | AK226505.1 | - | 6 - 7 | 904 | 99 | |
AY086218.1 | - | 6 - 7 | 932 | 99 | ||
AY150419.1 | - | 5 - 6 | 814 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |