Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128268.3 | NP_565635.1 | 7 - 8 | 1032 | 89 | |
GenBank mRNA | AF311221.1 | - | 7 - 8 | 866 | 89 | |
AF428305.1 | - | 7 - 8 | 871 | 89 | ||
AY039915.1 | - | 7 - 8 | 878 | 89 | ||
AY142569.1 | - | 7 - 8 | 842 | 89 | ||
AY143937.1 | - | 7 - 8 | 842 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |