Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCC8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203015.2 | NP_001189944.1 | 9 - 10 | 4346 | 123 | |
NM_001338517.1 | NP_001326627.1 | 10 - 11 | 4257 | 123 | ||
NM_001338518.1 | NP_001326629.1 | 9 - 10 | 4236 | 123 | ||
NM_001338519.1 | NP_001326630.1 | 9 - 10 | 4084 | 123 | ||
NM_001338520.1 | NP_001326628.1 | 10 - 11 | 4028 | 123 | ||
NM_113020.3 | NP_188762.3 | 9 - 10 | 4194 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |