Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PMP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113321.5 | NP_189058.1 | 6 - 7 | 655 | 75 | |
GenBank mRNA | AF104329.1 | - | 6 - 7 | 519 | 75 | |
AY053413.1 | - | 6 - 7 | 535 | 75 | ||
AY085022.1 | - | 6 - 7 | 555 | 75 | ||
AY133587.1 | - | 6 - 7 | 470 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |